Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: MAAESLPFS FGTLSSWELE AWYEDLQEVL SSDENGGTYV SPPGNEEEES KIFTTLDPAS LAWLTEEEPE PAEVTSTSQS PHSPDSSQSS LAQEEEEEDQ GRTRKRKQSG HSPARAGKQR MKEKEQENER KVAQLAEENE RLKQEIERLT REVEATRRAL IDRMVNLHQA (1-169 aa encoded by BC003637)
GADD153 is a small nuclear protein that is capable of dimerizing with transcription factors C/EBP alpha and beta. Once dimerized, this complex inhibits the normal binding and function of C/EBP to classical binding sites. Inversely, the C/EBP GADD153 dimer gains binding activity to other non classical C/EBP stress related targets. Under normal cellular conditions this protein is not expressed in detectable levels, but is highly unregulated during times of cellular/ER stress. Examples of GADD153 inducing stress include: treatment with tunicamycin, nutrient starvation and reducing agents that interfere with the calcium flux across the ER membrane.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: alternative DDIT3 protein; alternative DDIT3 proteins; C/EBP homologous protein; C/EBP homoologous protein 10; C/EBP zeta; c/EBP-homologous protein 10; CCAAT/enhancer-binding protein homologous protein; chop-10; DDIT-3; growth arrest and DNA damage-inducible protein GADD153; growth arrest and DNA-damage-inducible protein GADD153; MGC4154; unnamed protein product
Gene Aliases: AltDDIT3; C/EBPzeta; CEBPZ; CHOP; CHOP-10; CHOP10; GADD153; RM4
UniProt ID: (Human) P35638, (Mouse) P35639
Entrez Gene ID: (Human) 1649, (Mouse) 13198, (Rat) 29467
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support