Percent identity with other species by BLAST analysis: Human, Gorilla, Orangutan, Gibbon, Monkey (100%).
The muscarinic cholinergic receptors belong to a larger family of G protein-coupled receptors. The functional diversity of these receptors is defined by the binding of acetylcholine and includes cellular responses such as adenylate cyclase inhibition, phosphoinositide degeneration, and potassium channel mediation. Muscarinic receptors influence many effects of acetylcholine in the central and peripheral nervous system. The muscarinic cholinergic receptor 3 controls smooth muscle contraction and its stimulation causes secretion of glandular tissue.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: acetylcholine receptor; acetylcholine receptor, muscarinic 3; cholinergic receptor; cholinergic receptor, muscarinic 3; m3 muscarinic receptor; Muscarinic acetylcholine receptor M3
Gene Aliases: CHRM3; EGBRS; HM3; PBS
UniProt ID: (Human) P20309
Entrez Gene ID: (Human) 1131
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support