Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 7
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
MA5-31689 has been tested in indirect ELISA.
After binding acetylcholine, the AChR responds by an extensive change in conformation that affects all subunits and leads to opening of an ion-conducting channel across the plasma membrane.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: acetylcholine receptor; acetylcholine receptor, nicotinic, alpha 6 (neuronal); alpha(alpha)6 nicotinic acetylcholine receptor (nAChR) subunit; cholinergic receptor; cholinergic receptor, nicotinic alpha 6; cholinergic receptor, nicotinic, alpha 6 (neuronal); cholinergic receptor, nicotinic, alpha polypeptide 6; unnamed protein product
Gene Aliases: CHNRA6
UniProt ID: (Human) Q15825
Entrez Gene ID: (Human) 8973
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support