Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • CHRNA7 Antibodies

          Invitrogen

          CHRNA7 Recombinant Rabbit Monoclonal Antibody (PSH19-20)

          Advanced Verification
          View all (19) CHRNA7 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite CHRNA7 Recombinant Rabbit Monoclonal Antibody (PSH19-20)

          Additional Information:

          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (11)
          • Advanced Verification (2)
          CHRNA7 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          CHRNA7 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 13

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          CHRNA7 Antibody (MA571395) in ICC/IF

          Immunofluorescence analysis of CHRNA7 using Jurkat. The cells were fixed in 4% paraformaldehyde for 15 minutes at room temperature, permeabilized with 0.1% Triton X-100 in PBS for 15 minutes at room temperature, then blocked with 1% BSA in 10% negative goat serum for 1 hour at room temperature. The cells were co-labeled with CHRNA7 Recombinant Rabbit Monoclonal Antibody (PSH19-20) (Product # MA571395) (green) at 1:500 dilution and beta tubulin (red) in 1% BSA in PBST overnight at 4 degrees Celsius and then with iFluor™ 488 Goat Anti-Rabbit... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          CHRNA7 Antibody in Immunocytochemistry (ICC/IF)
          CHRNA7 Antibody in Immunocytochemistry (ICC/IF)
          CHRNA7 Antibody in Immunocytochemistry (ICC/IF)
          CHRNA7 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          CHRNA7 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          CHRNA7 Antibody in Immunohistochemistry (Frozen) (IHC (F))
          CHRNA7 Antibody in Western Blot (WB)
          CHRNA7 Antibody in Western Blot (WB)
          CHRNA7 Antibody in Western Blot (WB)
          CHRNA7 Antibody in Flow Cytometry (Flow)
          CHRNA7 Antibody in Flow Cytometry (Flow)
          CHRNA7 Antibody
          CHRNA7 Antibody

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          CHRNA7 Recombinant Rabbit Monoclonal Antibody (PSH19-20)

          Product Details

          MA571395

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:5,000
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:1,000
          -

          Immunohistochemistry (Frozen) (IHC (F))

          1:500
          -

          Immunocytochemistry (ICC/IF)

          1:500
          -

          Flow Cytometry (Flow)

          1:1,000
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Expression System

          HEK293 cells

          Class

          Recombinant Monoclonal

          Type

          Antibody

          Clone

          PSH19-20

          Immunogen

          Recombinant protein within human CHRNA7 aa 290-502.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4, with 0.1% BSA, 40% glycerol

          Contains

          0.2% ProClin 950

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Positive Control: Jurkat cell lysate, HeLa cell lysate, A431 cell lysate, Caco-2 cell lysate, RAW264.7 cell lysate, C6 cell lysate, PC-12 cell lysate, Mouse brain tissue lysate, Mouse spleen tissue lysate, Rat brain tissue lysate, Rat spleen tissue lysate, mouse dorsal root ganglion tissue, rat dorsal root ganglion tissue, Jurkat, PC-12.

          Tissue Specificity: Tissue enhanced (adrenal gland, intestine).

          Subcellular Location: Postsynaptic cell membrane, cell membrane.

          Sequence Similarities: 91% Mouse/Rat

          Predicted band size: 56 kDa.

          Target Information

          Chromosome 15 encodes over 700 genes and comprises nearly 3% of the human genome. CHRFAM7A, a fusion protein comprised of FAM7A and AChR alpha7, is encoded by the CHRFAM7A gene, which is located on a region of human chromosome 15 that is associated with several neuropsychiatric disorders, such as schizophrenia and bipolar affective disorder. CHRFAM7A, also known as D-10 or CHRNA7, is a 412 amino acid multi-pass membrane protein that belongs to the nicotinic acetylcholine receptor (nAChR) subfamily of ligand-gated ion channel proteins, a protein superfamily that mediates signal transmission at synapses. CHRFAM7A is expressed in the hippocampus. Alternative splicing of the CHRFAM7A gene produces two splice variants.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: a7 nicotinic acetylcholine receptor; acetylcholine receptor; acetylcholine receptor alpha 7 neural; alpha 7 neuronal nicotinic acetylcholine receptor; alpha 7-2; alpha-7 nicotinic cholinergic receptor subunit; alpha7 nicotinic receptor; alpha7-nAChR; bungarotoxin alpha; C holinergic receptor, nicotinic, alpha polypeptide 7 (neuronal nicotinic acetycholine receptor alpha 7) (bungarotoxin alpha); cholinergic receptor; cholinergic receptor, nicotinic alpha 7; cholinergic receptor, nicotinic, alpha 7 (neuronal); cholinergic receptor, nicotinic, alpha polypeptide 7; CHRFAM7A; CHRNA7-DR1; cytotransmitter gated ion channel; D-10; nAChR alpha7; neuronal acetylcholine receptor protein, alpha-7 chain; neuronal nicotinic acetycholine receptor alpha 7; neurotransmitter receptor; acra7; nicotinic acetylcholine receptor subunit alpha-7; nicotinic receptor alpha 7 subunit; unnamed protein product

          View more View less

          Gene Aliases: a7nAChR; Acra7; alpha7; BTX; CHRNA7-2; nAchR; nAChR7; NACHRA7; NARAD

          View more View less

          UniProt ID: (Human) P36544, (Mouse) P49582, (Rat) Q05941

          View more View less

          Entrez Gene ID: (Human) 1139, (Mouse) 11441, (Rat) 25302

          View more View less

          Function(s)
          beta-amyloid binding transmembrane signaling receptor activity ion channel activity extracellular ligand-gated ion channel activity calcium channel activity protein binding acetylcholine receptor activity toxic substance binding chloride channel regulator activity acetylcholine-gated cation channel activity neurotransmitter receptor activity acetylcholine binding protein homodimerization activity adenylate cyclase binding protein kinase binding ATPase binding scaffold protein binding transmitter-gated ion channel activity involved in regulation of postsynaptic membrane potential
          Process(es)
          response to hypoxia positive regulation of heart rate involved in baroreceptor response to decreased systemic arterial blood pressure ion transport cation transport calcium ion transport cellular calcium ion homeostasis endocytosis signal transduction chemical synaptic transmission synaptic transmission, cholinergic learning or memory memory positive regulation of cell proliferation associative learning response to cold positive regulation of gene expression regulation of norepinephrine secretion neuronal action potential sperm motility response to food regulation of synaptic transmission, dopaminergic negative regulation of interleukin-1 beta production negative regulation of interleukin-6 production negative regulation of tumor necrosis factor production ion transmembrane transport response to nicotine behavioral response to nicotine T cell activation B cell activation regulation of membrane potential ovulation cycle negative regulation of I-kappaB kinase/NF-kappaB signaling positive regulation of MAPK cascade response to ethanol positive regulation of angiogenesis behavioral response to ethanol regulation of inflammatory response negative regulation of inflammatory response modulation of synaptic transmission synapse organization cognition sensory processing positive regulation of protein metabolic process regulation of synapse structural plasticity membrane depolarization excitatory postsynaptic potential synaptic transmission involved in micturition generation of ovulation cycle rhythm negative regulation of ERK1 and ERK2 cascade positive regulation of ERK1 and ERK2 cascade acetylcholine receptor signaling pathway dendritic spine organization modulation of excitatory postsynaptic potential presynaptic modulation of chemical synaptic transmission dendrite arborization negative regulation of cytokine production involved in inflammatory response positive regulation of long-term synaptic potentiation positive regulation of beta-amyloid formation negative regulation of beta-amyloid formation negative regulation of intracellular signal transduction regulation of amyloid precursor protein catabolic process response to amyloid-beta response to acetylcholine regulation of amyloid fibril formation activation of protein kinase C activity ligand-gated ion channel signaling pathway positive regulation of excitatory postsynaptic potential regulation of response to drug cell communication short-term memory signaling calcium ion transmembrane transport regulation of nitric oxide metabolic process response to carbamate
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-74d7b4b4d9-v4tl5:80/100.66.78.131:80.
          git-commit: 7aafa54bd143a040505a257d2a3ebda4095a617f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.56.0-Offline