Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Recommended positive controls: MCF-7.
Predicted reactivity: Cow: 92%; Dog: 100%; Guinea Pig: 92%; Mouse: 85%; Rabbit: 100%; Rat: 85%.
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
After binding acetylcholine, the AChR responds by an extensive change in conformation that affects all subunits and leads to opening of an ion-conducting channel across the plasma membrane.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: acetylcholine receptor; Acetylcholine receptor subunit epsilon; acetylcholine receptor, nicotinic, epsilon (muscle); AchR epsilon subunit; ACHRG; cholinergic receptor; cholinergic receptor, nicotinic epsilon; cholinergic receptor, nicotinic, epsilon (muscle); cholinergic receptor, nicotinic, epsilon polypeptide; MGC133376
Gene Aliases: ACHRE; CHRNE
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support