The function of this gene has not been reported. This gene contains a trinucleotide (ctt) repeat length polymorphism; alleles with either one or two repeat units have been identified.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CDK2-interacting protein; cyclin-dependent kinase 2 interacting protein; Cyclin-dependent kinase 2-interacting protein; MGC849
Gene Aliases: CINP
UniProt ID: (Human) Q9BW66
Entrez Gene ID: (Human) 51550
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support