    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • 360° CDMO and CRO Solutions
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
          • Primary Antibodies ›
          • SARS-CoV-2 Spike Protein RBD Antibodies
          Proteintech
          spike protein Recombinant Rabbit Monoclonal Antibody (4I18), CoraLite® Plus 750
          View all (118) SARS-CoV-2 Spike Protein RBD antibodies
          Tech Support
          Cite spike protein Recombinant Rabbit Monoclonal Antibody (4I18), CoraLite® Plus 750
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          spike protein Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          spike protein Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          spike protein Antibody (CL750-82734-4) in WB
          Various lysates were subjected to SDS PAGE followed by western blot with CL750-82805-2 (GRP94 antibody) at dilution of 1:1,000 incubated at room temperature for 1.5 hours. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          spike protein Antibody in Western Blot (WB)
          Product Details
          CL750-82734-4
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:500-1:2,000
          -
          Product Specifications
          Species Reactivity
          Virus
          Host/Isotype
          Rabbit / IgG
          Expression System
          CHO cells
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          4I18
          Immunogen
          spike protein fusion protein Ag30684
          Conjugate
          CoraLite® Plus 750 CoraLite® Plus 750 CoraLite® Plus 750
          • Unconjugated
          • Request custom conjugation
          Excitation/Emission Max
          755/780 nm
          Form
          Liquid
          Concentration
          1000 µg/mL
          Purification
          Affinity chromatography
          Storage buffer
          PBS, pH 7.3, with 50% glycerol, 0.5% BSA
          Contains
          0.05% ProClin 300
          Storage conditions
          -20°C, store in dark
          Shipping conditions
          Wet ice
          Product Specific Information
          Sequence of this protein is as follows: DFTGCVIAWN SNNLDSKVG DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN DFTGCVIAWN
          Target Information
          SARS-CoV-2 (Severe Acute Respiratory Syndrome Coronavirus 2) also known as 2019-nCoV (2019 Novel Coronavirus) is a virus that causes illnesses ranging from the common cold symptoms to severe effects including death and respiratory distress. The CoV Spike (S) protein plays the most important roles in viral attachment, fusion and entry, and serves as a target for development of antibodies, entry inhibitors and vaccines. Of note is the Spike protein receptor-binding domain (RBD, S-RBD) in SARS-CoV-2 Spike protein which binds strongly to human and bat angiotensin-converting enzyme 2 (ACE2) receptors. SARS-CoV-2 RBD exhibits significantly higher binding affinity to ACE2 receptor than SARS-CoV RBD in comparison. S1 contains a receptor-binding domain (RBD) that can specifically bind to angiotensin-converting enzyme 2 (ACE2), the receptor on target cells.
          • Technical Support Centers
          • Find Documents and Certificates
          Brasil flag icon
          Brasil
          Your items have has been added!
          Host server : magellan-search-green-7d94cb4b65-jwmpg:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0