Alzheimer's disease (AD) is characterized by deposition of beta-amyloid as senile plaques and cerebral amyloid angiopathy. The principal component of AD amyloid is the amyloid beta peptide (A beta). Although production and deposition of A beta appear closely related to the pathogenesis of AD, current evidence suggest that A beta alone is not sufficient to cause neuronal death and symptoms of dementia. A component of senile plaque (SP), CLAC (collagen-like Alzheimer amyloid plaque component ) and its precursor CLAC-P are deposited with extracellular beta-amyloid and have been recently implicated in cell toxicity of beta-amyloid in AD brains. CLAC-P is a unique, membrane-bound collagen-like structure containing three Gly-X-Y repeat motifs and may define a novel class of neuronal collagens.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Alzheimer disease amyloid-associated protein; AMY; CLAC; CLAC-P; Collagen alpha-1(XXV) chain; Collagen-like Alzheimer amyloid plaque component; collagenous Alzheimer amyloid plaque component
Gene Aliases: AMY; CFEOM5; CLAC; CLAC-P; CLACP; COL25A1
UniProt ID: (Human) Q9BXS0
Entrez Gene ID: (Human) 84570
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support