Invitrogen
Product is shipped at room temperature as a lyophilized powder and should be stored at -20 °C upon receipt. Reconstitution: add 50 µL of deionized water.
This gene is a member of the CLC family of voltage-gated chloride channels. The encoded protein is predicted to have 12 transmembrane domains, and requires a beta subunit called barttin to form a functional channel. It is thought to function in salt reabsorption in the kidney and potassium recycling in the inner ear. The gene is highly similar to CLCNKB, which is located 10 kb downstream from this gene. Multiple transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: chloride channel K1; chloride channel K1-like; putative basolateral cTAL chloride channel ClC-Ka; chloride channel K2; Chloride channel Ka; Chloride channel Kb; Chloride channel protein ClC-Ka; Chloride channel protein ClC-Kb; ClC-K1; ClC-K2; CLCNK; hClC-Kb; putative basolateral cTAL chloride channel ClC-Ka; putative basolateral mTAL chloride channel ClC-Ka
Gene Aliases: C75963; CLC-K1; ClC-K2L; Clc-Ka; Clck2; CLCKB; Clcnk1; Clcnk1l; Clcnka; Clcnkb
UniProt ID: (Mouse) Q9WUB7, (Rat) Q06393, (Mouse) Q9WUB6, (Rat) P51802
Entrez Gene ID: (Mouse) 12733, (Rat) 79425, (Mouse) 56365, (Rat) 79430
Molecular Function:
anion channel
ion channel
transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support