The recommended shelf life for this product is 1 year from date of receipt.
The protein encoded by this gene belongs to the family of cytoplasmic linker proteins, which have been proposed to mediate the interaction between specific membranous organelles and microtubules. This protein was found to associate with both microtubules and an organelle called the dendritic lamellar body. This gene is hemizygously deleted in Williams syndrome, a multisystem developmental disorder caused by the deletion of contiguous genes at 7q11.23. Alternative splicing of this gene generates 2 transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CAP-Gly domain-containing linker protein 2; CLIP-115; cytoplasmic linker 2; Cytoplasmic linker protein 115; Cytoplasmic linker protein 2; testicular tissue protein Li 40; Williams-Beuren syndrome chromosomal region 3 protein; Williams-Beuren syndrome chromosomal region 4 protein; Williams-Beuren syndrome chromosome region 3; Williams-Beuren syndrome chromosome region 4
Gene Aliases: CLIP; CLIP-115; CLIP2; CYLN2; KIAA0291; WBSCR3; WBSCR4; WSCR3; WSCR4
UniProt ID: (Human) Q9UDT6
Entrez Gene ID: (Human) 7461
Molecular Function:
chaperone
cytoskeletal protein
microtubule family cytoskeletal protein
non-motor microtubule binding protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support