Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
For reconstitution, we recommend adding 100 µL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiments, we strongly recommend performing another round of desalting. (Zeba Spin Desalting Columns, 7KMWCO, 0.5 mL, Product # 89882)
CLIP4 includes 4 isoforms of the human protein that are produced by alternative splicing.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CAP-GLY domain containing linker protein family, member 4; CAP-Gly domain-containing linker protein 4; restin-like 2; Restin-like protein 2
Gene Aliases: 1700024K14Rik; 1700074B05Rik; 4833417L20Rik; 5830409B12Rik; CLIP4; RGD1311049; RSNL2
UniProt ID: (Human) Q8N3C7, (Mouse) Q8CI96, (Rat) Q66HD5
Entrez Gene ID: (Human) 79745, (Mouse) 78785, (Rat) 298801
Molecular Function: chaperone
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support