Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide Sequence: YPSRREPPPR RSRSRSHDRL PYQRRYRERR DSDTYRCEER SPSFGEDYYG PSRSRHRRRS RERGPYRTRK HAHHCHKRRT RSCSSASSRS QQSSKRSSRS V
CLK3 is a dual specificity kinase that can interact with and cause the nuclear redistribution of SR proteins. Two transcript variants encoding different isoforms have been found for CLK3. Related pseudogenes are located on chromosomes 1 and 9.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CDC-like kinase 3; Dual specificity protein kinase CLK3; Protein kinase CLK3
Gene Aliases: CLK3; PHCLK3; PHCLK3/152
UniProt ID: (Human) P49761, (Rat) Q63117
Entrez Gene ID: (Human) 1198, (Rat) 171305
Molecular Function: non-receptor serine/threonine protein kinase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support