Applications | Tested Dilution | Publications |
---|---|---|
Western Blot (WB) |
1:200 | - |
Product Specifications | |
---|---|
Species Reactivity |
Rat |
Host/Isotype |
Rabbit / IgG |
Class |
Polyclonal |
Type |
Antibody |
Immunogen |
Peptide corresponding to amino acid residues 41-54 of rat CNGA1 (Intracellular, N-terminus) |
Conjugate |
Unconjugated |
Form |
Lyophilized |
Concentration |
0.8 mg/mL |
Purification |
Antigen affinity chromatography |
Storage buffer |
PBS, pH 7.4, with 1% BSA |
Contains |
0.05% sodium azide |
Storage conditions |
-20°C |
RRID |
AB_2857290 |
Reconstitute antibody with 50 µL of double distilled water (DDW). The reconstituted solution can be stored at 4 degrees C for up to 1 week. For longer periods, small aliquots should be stored at -20 degrees C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 x g 5 min).
Peptide sequence is: (C)NDNGSLS EESENED
Homology: Mouse - 13/14 amino acid residues identical; human - 9/14 amino acid residues identical.
The protein encoded by this gene is involved in phototransduction. Along with another protein, the encoded protein forms a cGMP-gated cation channel in the plasma membrane, allowing depolarization of rod photoreceptors. This represents the last step in the phototransduction pathway. Defects in this gene are a cause of retinitis pigmentosa autosomal recessive (ARRP) disease. Two transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cGMP-gated cation channel alpha-1; cng channel; CNG channel alpha-1; CNG-1; CNG1; Cyclic nucleotide-gated cation channel 1; Cyclic nucleotide-gated channel alpha-1; Cyclic nucleotide-gated channel, photoreceptor; Rod photoreceptor cGMP-gated channel subunit alpha
Gene Aliases: Cncg; Cncg1; Cnga1; HCN
UniProt ID: (Rat) Q62927
Entrez Gene ID: (Rat) 85259
Molecular Function:
ligand-gated ion channel
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support