Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
This is a rabbit polyclonal antibody raised against a region from the C-terminal CNOT1 of human origin. CNOT1 exists as many isoforms. Isoform one is about 267 kDa and isoform four is about 173 kDa.
Immunogen sequence: MNLDSLSLA LSQISYLVDN LTKKNYRASQ QEIQHIVNRH GPEADRHLLR CLFSHVDFSG DGKSSGKDFH QTQFLIQECA LLITKPNFIS TLSYAIDNPL HYQKSLKPAP HLFAQLSKVL KLSKVQEVIF GLALLNSSSS DLRGFAAQFI KQKLPDLLRS YIDADVSGNQ EGGFQDIAIE VLHLLLSHLL FGQKGAFGVG QEQIDAFLKT LRRDFPQERC PVVLAPLLYP EKRDILMDRI LPDSGGVAKT MMESSLADFM QEVGYGFCAS IEECRNIIVQ FGVREVTAAQ VARVLGMMAR THSGLTDGIP LQSISAPGSG IWSDGKDKSD GAQAHTWNVE VLIDVLKELN PSLNF (1-354 aa encoded by BC040523)
Scaffolding component of the CCR4-NOT complex which is one of the major cellular mRNA deadenylases and is linked to various cellular processes including bulk mRNA degradation, miRNA-mediated repression, translational repression during translational initiation and general transcription regulation. Additional complex functions may be a consequence of its influence on mRNA expression. Its scaffolding function implies its interaction with the catalytic complex module and diverse RNA-binding proteins mediating the complex recruitment to selected mRNA 3'UTRs. Involved in degradation of AU-rich element (ARE)-containing mRNAs probably via association with ZFP36. Mediates the recruitment of the CCR4-NOT complex to miRNA targets and to the RISC complex via association with TNRC6A, TNRC6B or TNRC6C. Acts as a transcriptional repressor. Represses the ligand-dependent transcriptional activation by nuclear receptors. Involved in the maintenance of embryonic stem (ES) cell identity. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: adrenal gland protein AD-005; CCR4-associated factor 1; negative regulator of transcription subunit 1 homolog; NOT1 (negative regulator of transcription 1, yeast) homolog; unnamed protein product
Gene Aliases: 6030411K04Rik; AD-005; CDC39; D830048B13; HPE12; NOT1; NOT1H; RGD1308009; VIBOS
UniProt ID: (Human) A5YKK6, (Mouse) Q6ZQ08
Entrez Gene ID: (Pig) 100511993, (Human) 23019, (Rat) 291841, (Mouse) 234594
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support