Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide Sequence: PRWTSTQTSG PGLPIGFKGW NGQIFKVNTL LEPKVLSDDC KIQDGSWHKA TFLFHTQEPN QLPVIEVQKL PHLKTERKYY IDSSSVCFL
May participate in regulating type I collagen fibrillogenesis at specific anatomical locations during fetal development.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Collagen alpha-1(XXIV) chain; collagen, type XXIV, alpha 1
Gene Aliases: COL24A1
UniProt ID: (Human) Q17RW2
Entrez Gene ID: (Human) 255631
Molecular Function: extracellular matrix structural protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support