Product References
Antagonizing pathways leading to differential dynamics in colon carcinogenesis in Shugoshin1 (Sgo1)-haploinsufficient chromosome instability model.
Molecular carcinogenesis
Rao CV,Sanghera S,Zhang Y,Biddick L,Reddy A,Lightfoot S,Dai W,Yamada HY
Published figure using COX2 polyclonal antibody (Product # PA5-16817) in Immunohistochemistry
Sun May 01 00:00:00 EDT 2016
Aspirin blocks growth of breast tumor cells and tumor-initiating cells and induces reprogramming factors of mesenchymal to epithelial transition.
Laboratory investigation; a journal of technical methods and pathology
Maity G,De A,Das A,Banerjee S,Sarkar S,Banerjee SK
PA5-16817 was used in western blot to evaluate the effect of aspirin in mesenchymal to epithelial transition
Wed Jul 01 00:00:00 EDT 2015
PharmDB-K: Integrated Bio-Pharmacological Network Database for Traditional Korean Medicine.
PloS one
Lee JH,Park KM,Han DJ,Bang NY,Kim DH,Na H,Lim S,Kim TB,Kim DG,Kim HJ,Chung Y,Sung SH,Surh YJ,Kim S,Han BW
PA5-16817 was used in western blot to present PharmDB-K, a database offering comprehensive information relating to TKM-associated drugs, disease indication, and protein relationships
Mon Jun 13 00:00:00 EDT 2016
Genistein improves 3-NPA-induced memory impairment in ovariectomized rats: impact of its antioxidant, anti-inflammatory and acetylcholinesterase modulatory properties.
PloS one
Menze ET,Esmat A,Tadros MG,Abdel-Naim AB,Khalifa AE
PA5-16817 was used in immunohistochemistry - paraffin section to characterize how 3-NPA-induced memory impairment in ovariectomized rats is improved by Genistein and its anti-inflammatory, antioxidant, and acetylcholinesterase modulatory properties
Fri Nov 20 00:00:00 EST 2015
Comparative immunohistochemical assessment of cutaneous cyclooxygenase-2 enzyme expression in chronological aging and photoaging.
Photodermatology, photoimmunology & photomedicine
Habib MA,Salem SA,Hakim SA,Shalan YA
PA5-16817 was used in immunohistochemistry to perform a comparative study of the immunohistochemical expression of COX-2 in skin samples from individuals with chronologically or photo aged skin
Sat Feb 01 00:00:00 EST 2014
Immunohistochemical evaluation of COX-1 and COX-2 expression in keloid and hypertrophic scar.
The American Journal of dermatopathology
Abdou AG,Maraee AH,Saif HF
PA5-16817 was used in immunohistochemistry to evaluate COX-1 and COX-2 expression in keloid and hypertrophic scars
Tue Apr 01 00:00:00 EDT 2014
Therapeutic effect of coenzyme Q10 against experimentally-induced hepatocellular carcinoma in rats.
Environmental toxicology and pharmacology
Fouad AA,Al-Mulhim AS,Jresat I
PA5-16817 was used in immunohistochemistry to study the therapeutic effects of coenzyme Q10 in a rat model of hepatocellular carcinoma
Tue Jan 01 00:00:00 EST 2013
Simvastatin treatment ameliorates injury of rat testes induced by cadmium toxicity.
Biological trace element research
Fouad AA,Albuali WH,Jresat I
PA5-16817 was used in immunohistochemistry to study the mechanisms underlying the protection against cadmium-induced testicular toxicity conferred by treatment with simvastatin
Sat Jun 01 00:00:00 EDT 2013
The effects of ulipristal on Bax/Bcl-2, cytochrome c, Ki-67 and cyclooxygenase-2 expression in a rat model with surgically induced endometriosis.
European journal of obstetrics, gynecology, and reproductive biology
Huniadi CA,Pop OL,Antal TA,Stamatian F
PA5-16817 was used in immunohistochemistry to use a rat model of endometriosis to study the effects of ulipristal on endometrial histology and the expression of Bax/Bcl-2, cytochrome c, Ki-67 and COX-2
Mon Jul 01 00:00:00 EDT 2013
A promise in the treatment of endometriosis: an observational cohort study on ovarian endometrioma reduction by N-acetylcysteine.
Evidence-based complementary and alternative medicine : eCAM
Porpora MG,Brunelli R,Costa G,Imperiale L,Krasnowska EK,Lundeberg T,Nofroni I,Piccioni MG,Pittaluga E,Ticino A,Parasassi T
PA5-16817 was used in immunohistochemistry to study the efficacy of N-acetylcysteine therapy in women with ovarian endometriosis
Thu Jul 04 00:00:00 EDT 2013
Reappraisal of the therapeutic role of celecoxib in cholangiocarcinoma.
PloS one
Yeh CN,Chiang KC,Juang HH,S Pang JH,Yu CS,Lin KJ,Yeh TS,Jan YY
PA5-16817 was used in immunohistochemistry to evaluate the efficacy and safety of celecoxib to treat cholangiocarcinoma
Tue Apr 29 00:00:00 EDT 2014
Suppression of β-catenin and cyclooxygenase-2 expression and cell proliferation in azoxymethane-induced colonic cancer in rats by rice bran phytic acid (PA).
Asian Pacific journal of cancer prevention : APJCP
Saad N,Esa NM,Ithnin H
PA5-16817 was used in immunohistochemistry to study the ability of phytic acid from rice bran to reduce the expression of beta-catenin and COX-2 and to inhibit colon cancer proliferation in a rat carcinogen-induced model
Tue Oct 14 00:00:00 EDT 2014
Multimechanistic antifibrotic effect of biochanin a in rats: implications of proinflammatory and profibrogenic mediators.
PloS one
Breikaa RM,Algandaby MM,El-Demerdash E,Abdel-Naim AB
PA5-16817 was used in immunohistochemistry to assess the antifibrotic of biochanin A
Wed Feb 19 00:00:00 EST 2014
Protective effects of nebivolol against cold restraint stress-induced gastric ulcer in rats: role of NO, HO-1, and COX-1,2.
Nitric oxide : biology and chemistry
Morsy MA,Heeba GH,Abdelwahab SA,Rofaeil RR
PA5-16817 was used in immunohistochemistry to study the protective mechanism of nebivolol in a rat gastric ulcer model
Wed Aug 15 00:00:00 EDT 2012
Role of immunohistochemical cyclo-oxygenase-2 (COX-2) and osteocalcin in differentiating between osteoblastomas and osteosarcomas.
The Malaysian journal of pathology
El-Badawi ZH,Muhammad EM,Noaman HH
PA5-16817 was used in immunohistochemistry to study the differentiation of osteoblastomas and osteosarcomas based on the immunohistochemical measurement of COX-2 and osteocalcin
Fri Jun 01 00:00:00 EDT 2012
Cyclooxygenase-2 expression and its prognostic significance in clear cell renal cell carcinoma.
Korean journal of pathology
Lee JW,Park JH,Suh JH,Nam KH,Choe JY,Jung HY,Chae JY,Moon KC
PA5-16817 was used in immunohistochemistry and western blot to study the prognostic value of the immunohistochemical expression of COX-2 in patients with clear cell renal cell carcinoma
Fri Jun 01 00:00:00 EDT 2012
Inhibition of COX-2 in colon cancer modulates tumor growth and MDR-1 expression to enhance tumor regression in therapy-refractory cancers in vivo.
Neoplasia (New York, N.Y.)
Rahman M,Selvarajan K,Hasan MR,Chan AP,Jin C,Kim J,Chan SK,Le ND,Kim YB,Tai IT
PA5-16817 was used in immunohistochemistry and western blot to study the beneficial effects on tumor growth and MDR-1 expression of pharmacological COX-2 inhibition in chemoresistant colorectal cancer
Sun Jul 01 00:00:00 EDT 2012
Resveratrol suppresses 4-hydroxyestradiol-induced transformation of human breast epithelial cells by blocking IκB kinaseβ-NF-κB signalling.
Free radical research
Park SA,Na HK,Surh YJ
PA5-16817 was used in western blot to study the role of IKK-beta and NFkB in the mechanism by which resveratrol blocks human breast epithelial transformation induced by 4-hydroxyestradiol
Wed Aug 01 00:00:00 EDT 2012
The acid sphingomyelinase inhibitors block interferon-α-induced serotonin uptake via a COX-2/Akt/ERK/STAT-dependent pathway in T cells.
International immunopharmacology
Su HC,Ma CT,Lin CF,Wu HT,Chuang YH,Chen LJ,Tsao CW
PA5-16817 was used in western blot to study the role of sphingomyelinase inhibitors in serotonin uptake
Tue Nov 01 00:00:00 EDT 2011
Cox-2 deletion in myeloid and endothelial cells, but not in epithelial cells, exacerbates murine colitis.
Carcinogenesis
Ishikawa TO,Oshima M,Herschman HR
PA5-16817 was used in immunohistochemistry to investigate the involvement of COX2 expression in different cell types in the pathogenesis of colitis
Tue Mar 01 00:00:00 EST 2011
Cyclooxygenase (COX)-1 activity precedes the COX-2 induction in Aβ-induced neuroinflammation.
Journal of molecular neuroscience : MN
Dargahi L,Nasiraei-Moghadam S,Abdi A,Khalaj L,Moradi F,Ahmadiani A
PA5-16817 was used in western blot to investigate the activation of cyclooxygenases in the neuroinflammation mediated by Abeta
Thu Sep 01 00:00:00 EDT 2011
Tumor formation in a mouse model of colitis-associated colon cancer does not require COX-1 or COX-2 expression.
Carcinogenesis
Ishikawa TO,Herschman HR
PA5-16817 was used in immunohistochemistry to investigate the influence of COX-1 or COX-2 on specific tumor formation in mice
Thu Apr 01 00:00:00 EDT 2010