Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MLDVEAQEPP KGKWSTPPFD PRFPSQNQIR NCYQNFLDYH RCLKTRTRRG KSTQPCEYYF RVYHSLCPIS WVESWNEQIK NGIFAGKI
The cytochrome c oxidase (COX) family of proteins function as the final electron donor in the respiratory chain to drive a proton gradient across the inner mitochondrial membrane, ultimately resulting in the production of water. The mammalian COX apoenzyme is a dimer, with each monomer consisting of 13 subunits, some of which are mitochondrial and some of which are nuclear. Localized to the intermembrane space, COX6b2 (Cytochrome c oxidase subunit 6B2), also known as Cytochrome c oxidase subunit VIb isoform 2 and Cancer/testis antigen 59, is a 88 amino acid mitochondrial protein that is responsible for joining the two COX monomers to form the COX dimer. COX6b2 is specifically expressed in testis and is found to be upregulated in certain cancer cell lines.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: COX VIb-2; Cytochrome c oxidase subunit 6B2; Cytochrome c oxidase subunit VIb isoform 2; Cytochrome c oxidase subunit VIb, testis-specific isoform
Gene Aliases: 1700067P11Rik; BC048670; Cox6b2; COXVIB2
UniProt ID: (Mouse) Q80ZN9
Entrez Gene ID: (Mouse) 333182
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support