Predicted to react with Human samples.
Probable carboxypeptidase which may cleave proteins with C-terminal acidic residues.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Carboxypeptidase O; CPO; metallocarboxypeptidase C; metallocarboxypeptidase O
Gene Aliases: CPO
UniProt ID: (Human) Q8IVL8
Entrez Gene ID: (Human) 130749
Molecular Function:
hydrolase
metalloprotease
protease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support