Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The recommended shelf life for this product is 1 year from date of receipt.
Based on 100% sequence identity, this antibody is predicted to react with Mouse and Rat
FLJ12529 is a protein coding gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CFIm59; cleavage and polyadenylation specific factor 7, 59kDa; Cleavage and polyadenylation specificity factor 59 kDa subunit; Cleavage and polyadenylation specificity factor subunit 7; Cleavage factor Im complex 59 kDa subunit; CPSF 59 kDa subunit; FLJ12529; pre mRNA cleavage factor I, 59 kDa subunit; pre-mRNA cleavage factor I, 59 kDa subunit; Pre-mRNA cleavage factor Im 59 kDa subunit
Gene Aliases: CFIm59; CPSF7
UniProt ID: (Human) Q8N684
Entrez Gene ID: (Human) 79869
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support