Invitrogen
Applications | Tested Dilution | Publications |
---|---|---|
Flow Cytometry (Flow) |
Not Cited | - |
Product Specifications | |
---|---|
Species Reactivity |
Human |
Host/Isotype |
Rabbit / IgG |
Class |
Recombinant Monoclonal |
Type |
Antibody |
Clone |
004 |
Immunogen |
Recombinant Human CRABP2 protein |
Conjugate |
FITC |
Excitation/Emission Profile |
View spectra |
Form |
Liquid |
Concentration |
0.1 mg/mL |
Storage conditions |
4° C, store in dark, DO NOT FREEZE! |
May be stored for twelve months without detectable loss of activity. Protect from prolonged exposure to light and do not freeze. Sodium azide is toxic to cells and should be disposed of properly so flush with large volumes of water during disposal.
Cellular retinoic acid binding protein (CRABP) is a low molecular weight protein with unknown function and existing in two isoforms, CRABP1 and CRABP2. The predicted amino acid sequences of human CRABP1 and CRABP2 exhibits 99.3 and 93.5% identity to mouse CRABP1 and CRABP2, respectively. The CRABP2 isoform is important in retinoic acid-mediated regulation of human skin growth and differentiation. The CRABP2 gene localized on human chromosome 1q21.3 is transcriptionally regulated by a newly synthesized regulatory protein. The protein is expressed in human adult liver.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.
Protein Aliases: Cellular retinoic acid-binding protein 2; Cellular retinoic acid-binding protein II; CRABP-II; RP11-66D17.4
Gene Aliases: CRABP2
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support