Applications | Tested Dilution | Publications |
---|---|---|
Western Blot (WB) |
1:500-1:2,000 | - |
Immunohistochemistry (Paraffin) (IHC (P)) |
1:50-1:500 | - |
Product Specifications | |
---|---|
Species Reactivity |
Human, Mouse, Pig |
Host/Isotype |
Mouse / IgG1 |
Class |
Monoclonal |
Type |
Antibody |
Clone |
2A9D11 |
Immunogen |
CRBN Fusion Protein Ag17166 (143-442 aa encoded by BC017419) |
Conjugate |
Unconjugated |
Form |
Liquid |
Concentration |
1 mg/mL |
Purification |
Protein G |
Storage buffer |
PBS, pH 7.3, with 50% glycerol |
Contains |
0.02% sodium azide |
Storage conditions |
-20°C |
Immunogen sequence: AYREEQDFG IEIVKVKAIG RQRFKVLELR TQSDGIQQAK VQILPECVLP STMSAVQLES LNKCQIFPSK PVSREDQCSY KWWQKYQKRK FHCANLTSWP RWLYSLYDAE TLMDRIKKQL REWDENLKDD SLPSNPIDFS YRVAACLPID DVLRIQLLKI GSAIQRLRCE LDIMNKCTSL CCKQCQETEI TTKNEIFSLS LCGPMAAYVN PHGYVHETLT VYKACNLNLI GRPSTEHSWF PGYAWTVAQC KICASHIGWK FTATKKDMSP QKFWGLTRSA LLPTIPDTED EISPDKVILC L (143-442 aa encoded by BC017419)
CRBN, a member of the Lon protease protein family, plays a role in brain development. It is a component of the DCX (DDB1-CUL4-X-box) E3 protein ligase complex, a complex that mediates the ubiquitination and subsequent proteasomal degradation of target proteins and is required for limb outgrowth and expression of the fibroblast growth factor FGF8. CRBN is thought to regulate the assembly and neuronal surface expression of large-conductance calcium-activated potassium channels in brain regions involved in memory and learning via its interaction with KCNT1. It is widely expressed and highly expressed in brain.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DKFZp781K0715; MGC27358; novel lethal; Protein cereblon; Protein PiL; protein x 0001
Gene Aliases: 2610203G15Rik; 2900045O07Rik; AD-006; AF229032; AW108261; CRBN; MRT2; MRT2A; piL
UniProt ID: (Human) Q96SW2, (Mouse) Q8C7D2
Entrez Gene ID: (Pig) 100737786, (Human) 51185, (Mouse) 58799
Molecular Function:
ubiquitin-protein ligase
protein modifying enzyme
protease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support