Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
MA5-31709 has been tested in indirect ELISA.
The protein encoded by this gene is a component of the pre-mRNA-processing factor 19-cell division cycle 5-like (PRP19-CDC5L) protein complex, which activates pre-mRNA splicing and is an integral part of the spliceosome. The encoded protein is also a nuclear localization sequence binding protein, and binds to activation-induced deaminase and is important for antibody diversification. This gene may also be associated with the development of obesity. Alternative splicing results in multiple transcript variants. A pseudogene of this gene has been defined on the X chromosome. Diseases associated with CTNNBL1 include Immunodeficiency 99 With Hypogammaglobulinemia And Autoimmune Cytopenias.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Beta-catenin-like protein 1; catenin, beta like 1; NAP; nuclear associated protein; Nuclear-associated protein; RP5-1118M15.1; Testis development protein NYD-SP19
Gene Aliases: C20orf33; CTNNBL1; dJ633O20.1; NAP; P14L; PP8304
UniProt ID: (Human) Q8WYA6
Entrez Gene ID: (Human) 56259
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support