Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • CX3CR1/Albumin Antibodies

          Invitrogen

          CX3CR1/Albumin VHH-VHH-His Recombinant Alpaca Monoclonal Antibody (BI 655088)

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite CX3CR1/Albumin VHH-VHH-His Recombinant Alpaca Monoclonal Antibody (BI 655088)

          Additional Information:

          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}

          Product Details

          MA562791

          Applications
          Tested Dilution
          Publications

          ELISA (ELISA)

          Assay-dependent
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Alpaca / VHH

          Expression System

          CHO cells

          Class

          Recombinant Monoclonal

          Type

          Antibody

          Clone

          BI 655088

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Metal-chelate chromatography

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_3703890

          Target Information

          CX3CR1 also known as chemokine receptor 1 or fractalkine receptor is a G-protein-coupled receptor primarily expressed on subsets of immune cells such as monocytes natural killer cells and T lymphocytes. It binds to the chemokine ligand CX3CL1 (fractalkine) which is unique due to its membrane-bound nature and participates in critical processes like leukocyte adhesion and migration. This receptor-ligand interaction is vital for immune surveillance inflammation and tissue repair. Dysregulation of CX3CR1 is linked to various pathological states including chronic inflammatory diseases and cancer metastasis. In research contexts CX3CR1 is studied for its role in neuroinflammation and cardiovascular diseases where its modulation could offer therapeutic benefits. Albumin on the other hand is a versatile protein found in blood plasma and is the most abundant protein in human serum. It is crucial for maintaining osmotic pressure and acts as a carrier molecule for numerous endogenous and exogenous substances including hormones vitamins and drugs. Albumin's ability to bind and transport various molecules makes it an essential player in drug delivery systems and it is widely used in formulations to enhance the pharmacokinetics and bioavailability of therapeutic agents. The interaction between albumin and other drug molecules is a significant factor in pharmacology influencing drug distribution and clearance from the body.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: beta chemokine receptor-like 1; C-X3-C CKR-1; chemokine (C-C) receptor-like 1; chemokine (C-X3-C motif) receptor 1; chemokine (C-X3-C) receptor 1; Chemokine receptor 1; CMK-BRL-1; CMK-BRL1; fractalkine receptor; G-protein coupled receptor 13; Human albumin; Serum albumin; unnamed protein product

          View more View less

          Gene Aliases: CCRL1; CMKBRL1; CMKDR1; GPR13; GPRV28; V28

          View more View less

          UniProt ID: (Human) P49238

          View more View less

          Entrez Gene ID: (Human) 1524

          View more View less

          Function(s)
          cytokine receptor activity G-protein coupled receptor activity chemokine receptor activity protein binding G-protein coupled peptide receptor activity C-C chemokine receptor activity C-X3-C chemokine receptor activity C-C chemokine binding C-X3-C chemokine binding CX3C chemokine receptor binding
          Process(es)
          positive regulation of neuroblast proliferation adaptive immune response microglial cell activation involved in immune response response to ischemia chemotaxis immune response cellular defense response cell adhesion G-protein coupled receptor signaling pathway phospholipase C-activating G-protein coupled receptor signaling pathway positive regulation of cytosolic calcium ion concentration cell-cell signaling brain development response to wounding cytokine-mediated signaling pathway calcium-mediated signaling central nervous system maturation leukocyte chemotaxis regulation of tumor necrosis factor production negative regulation of interleukin-1 beta production social behavior autocrine signaling positive regulation of I-kappaB kinase/NF-kappaB signaling innate immune response regulation of nitric oxide biosynthetic process regulation of synaptic plasticity host-mediated regulation of intestinal microbiota composition regulation of neurogenesis positive regulation of neurogenesis modulation of synaptic transmission leukocyte tethering or rolling positive regulation of protein kinase B signaling synapse maturation cell chemotaxis antifungal innate immune response chemokine-mediated signaling pathway cellular response to lipopolysaccharide cellular response to cytokine stimulus positive regulation of monocyte chemotaxis synapse pruning negative regulation of hippocampal neuron apoptotic process multiple spine synapse organization, single dendrite regulation of microglial cell migration negative regulation of microglial cell mediated cytotoxicity negative regulation of apoptotic signaling pathway
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-74d7b4b4d9-v4tl5:80/100.66.78.131:80.
          git-commit: 7aafa54bd143a040505a257d2a3ebda4095a617f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.56.0-Offline