Product References
Surface functionalization of electrospun scaffolds using recombinant human decorin attracts circulating endothelial progenitor cells.
Scientific reports
Hinderer S,Sudrow K,Schneider M,Holeiter M,Layland SL,Seifert M,Schenke-Layland K
500-P87A was used in Immunofluorescence to demonstrate DCN as promising EPC-attracting coating, which is particularily interesting when aiming to generate off-the-shelf biomaterials with the potential of in vivo cell seeding.
Mon Jan 08 00:00:00 UTC 2018
Surface functionalization of electrospun scaffolds using recombinant human decorin attracts circulating endothelial progenitor cells.
Scientific reports
Hinderer S,Sudrow K,Schneider M,Holeiter M,Layland SL,Seifert M,Schenke-Layland K
500-P87A was used in Immunofluorescence to demonstrate DCN as promising EPC-attracting coating, which is particularily interesting when aiming to generate off-the-shelf biomaterials with the potential of in vivo cell seeding.
Mon Jan 08 00:00:00 UTC 2018
Chemokine axes in breast cancer: factors of the tumor microenvironment reshape the CCR7-driven metastatic spread of luminal-A breast tumors.
Journal of leukocyte biology
Weitzenfeld P,Kossover O,Körner C,Meshel T,Wiemann S,Seliktar D,Legler DF,Ben-Baruch A
500-P87A was used in Enzyme-linked immunosorbent assay to study the importance of chemokine axes in regulating the metastatic spread of breast cancer.
Wed Jun 01 00:00:00 UTC 2016
CXCL12 mediates trophic interactions between endothelial and tumor cells in glioblastoma.
PloS one
Rao S,Sengupta R,Choe EJ,Woerner BM,Jackson E,Sun T,Leonard J,Piwnica-Worms D,Rubin JB
Mon Aug 20 00:00:00 UTC 2012
The chemokine CXCL12 regulates monocyte-macrophage differentiation and RUNX3 expression.
Blood
Sánchez-Martín L,Estecha A,Samaniego R,Sánchez-Ramón S,Vega MÁ,Sánchez-Mateos P
Thu Jan 06 00:00:00 UTC 2011
CXCR7 influences leukocyte entry into the CNS parenchyma by controlling abluminal CXCL12 abundance during autoimmunity.
The Journal of experimental medicine
Cruz-Orengo L,Holman DW,Dorsey D,Zhou L,Zhang P,Wright M,McCandless EE,Patel JR,Luker GD,Littman DR,Russell JH,Klein RS
Mon Feb 14 00:00:00 UTC 2011
Intravenous administration of mesenchymal stem cells exerts therapeutic effects on parkinsonian model of rats: focusing on neuroprotective effects of stromal cell-derived factor-1alpha.
BMC neuroscience
Wang F,Yasuhara T,Shingo T,Kameda M,Tajiri N,Yuan WJ,Kondo A,Kadota T,Baba T,Tayra JT,Kikuchi Y,Miyoshi Y,Date I
Mon Apr 26 00:00:00 UTC 2010
IL-1R signaling within the central nervous system regulates CXCL12 expression at the blood-brain barrier and disease severity during experimental autoimmune encephalomyelitis.
Journal of immunology (Baltimore, Md. : 1950)
McCandless EE,Budde M,Lees JR,Dorsey D,Lyng E,Klein RS
Wed Jul 01 00:00:00 UTC 2009
Highly efficient differentiation of hESCs to functional hepatic endoderm requires ActivinA and Wnt3a signaling.
Proceedings of the National Academy of Sciences of the United States of America
Hay DC,Fletcher J,Payne C,Terrace JD,Gallagher RC,Snoeys J,Black JR,Wojtacha D,Samuel K,Hannoun Z,Pryde A,Filippi C,Currie IS,Forbes SJ,Ross JA,Newsome PN,Iredale JP
Tue Aug 26 00:00:00 UTC 2008
Blocking CXCR4-mediated cyclic AMP suppression inhibits brain tumor growth in vivo.
Cancer research
Yang L,Jackson E,Woerner BM,Perry A,Piwnica-Worms D,Rubin JB
Mon Jan 15 00:00:00 UTC 2007
Blocking CXCR4-mediated cyclic AMP suppression inhibits brain tumor growth in vivo.
Cancer research
Yang L,Jackson E,Woerner BM,Perry A,Piwnica-Worms D,Rubin JB
Mon Jan 15 00:00:00 UTC 2007
Shedding of syndecan-1 by stromal fibroblasts stimulates human breast cancer cell proliferation via FGF2 activation.
The Journal of biological chemistry
Su G,Blaine SA,Qiao D,Friedl A
Fri May 18 00:00:00 UTC 2007
CXCL12 limits inflammation by localizing mononuclear infiltrates to the perivascular space during experimental autoimmune encephalomyelitis.
Journal of immunology (Baltimore, Md. : 1950)
McCandless EE,Wang Q,Woerner BM,Harper JM,Klein RS
Fri Dec 01 00:00:00 UTC 2006
Identification of carboxypeptidase N as an enzyme responsible for C-terminal cleavage of stromal cell-derived factor-1alpha in the circulation.
Blood
Davis DA,Singer KE,De La Luz Sierra M,Narazaki M,Yang F,Fales HM,Yarchoan R,Tosato G
Wed Jun 15 00:00:00 UTC 2005
Chemokine receptor expression in EBV-associated lymphoproliferation in hu/SCID mice: implications for CXCL12/CXCR4 axis in lymphoma generation.
Blood
Piovan E,Tosello V,Indraccolo S,Cabrelle A,Baesso I,Trentin L,Zamarchi R,Tamamura H,Fujii N,Semenzato G,Chieco-Bianchi L,Amadori A
Tue Feb 01 00:00:00 UTC 2005
Differential processing of stromal-derived factor-1alpha and stromal-derived factor-1beta explains functional diversity.
Blood
De La Luz Sierra M,Yang F,Narazaki M,Salvucci O,Davis D,Yarchoan R,Zhang HH,Fales H,Tosato G
Thu Apr 01 00:00:00 UTC 2004
Selective expression of stromal-derived factor-1 in the capillary vascular endothelium plays a role in Kaposi sarcoma pathogenesis.
Blood
Yao L,Salvucci O,Cardones AR,Hwang ST,Aoki Y,De La Luz Sierra M,Sajewicz A,Pittaluga S,Yarchoan R,Tosato G
Mon Dec 01 00:00:00 UTC 2003
Regulation of endothelial cell branching morphogenesis by endogenous chemokine stromal-derived factor-1.
Blood
Salvucci O,Yao L,Villalba S,Sajewicz A,Pittaluga S,Tosato G
Mon Apr 15 00:00:00 UTC 2002
Regional expression of CXCL12/CXCR4 in liver and hepatocellular carcinoma and cell-cycle variation during in vitro differentiation.
Japanese journal of cancer research : Gann
Shibuta K,Mori M,Shimoda K,Inoue H,Mitra P,Barnard GF
Mon Jul 01 00:00:00 UTC 2002
Chemoattractants induce a rapid and transient upregulation of monocyte alpha4 integrin affinity for vascular cell adhesion molecule 1 which mediates arrest: an early step in the process of emigration.
The Journal of experimental medicine
Chan JR,Hyduk SJ,Cybulsky MI
Mon May 21 00:00:00 UTC 2001
Role of tyrosine phosphorylation in ligand-independent sequestration of CXCR4 in human primary monocytes-macrophages.
The Journal of biological chemistry
Wang J,Guan E,Roderiquez G,Calvert V,Alvarez R,Norcross MA
Fri Dec 28 00:00:00 UTC 2001