Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Predicted to react with Mouse and Rat samples.
CYB561D1 gene ontology (GO) annotations include oxidation-reduction process.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cytochrome b-561 domain containing 1; Cytochrome b561 domain-containing protein 1; cytochrome b561 family, member D1; Probable transmembrane reductase CYB561D1
Gene Aliases: 1600010M23Rik; AI605489; AW457162; CYB561D1
UniProt ID: (Human) Q8N8Q1, (Mouse) A2AE42
Entrez Gene ID: (Human) 284613, (Rat) 362017, (Mouse) 72023
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support