Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The recommended shelf life for this product is 1 year from date of receipt.
Component of the CYFIP1-EIF4E-FMR1 complex which binds to the mRNA cap and mediates translational repression. In the CYFIP1-EIF4E-FMR1 complex this subunit is an adapter between EIF4E and FMR1. Promotes the translation repression activity of FMR1 in brain probably by mediating its association with EIF4E and mRNA. Involved in formation of membrane ruffles and lamellipodia protrusions and in axon outgrowth. Binds to F-actin but not to RNA. Regulator of epithelial morphogenesis. May act as an invasion suppressor in cancers.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CYFIP1; Cytoplasmic FMR1-interacting protein 1; cytoplasmic FMRP interacting protein 1; FLJ45151; p140sra-1; selective hybridizing clone; Specifically Rac1-associated protein 1; Sra-1
Gene Aliases: CYFIP1; KIAA0068; P140SRA-1; SHYC; SRA-1; SRA1
UniProt ID: (Human) Q7L576
Entrez Gene ID: (Human) 23191
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support