Product is shipped at room temperature as a lyophilized powder and should be stored at -20 °C upon receipt. Reconstitution: add 50 µL of deionized water.
Receptor for cysteinyl leukotrienes. The response is mediated via a G-protein that activates a phosphatidylinositol-calcium second messenger system. Stimulation by BAY u9773, a partial agonist, induces specific contractions of pulmonary veins and might also have an indirect role in the relaxation of the pulmonary vascular endothelium. The rank order of affinities for the leukotrienes is LTC4 = LTD4 >> LTE4.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CysLT(2); CysLT-2; CysLTR2; cysteinyl leukotriene CysLT2 receptor; Cysteinyl leukotriene receptor 2; G-protein coupled receptor GPCR21; G-protein coupled receptor HG57; hGPCR21; HPN321
Gene Aliases: CYSLT2; CYSLT2R; CYSLTR2; GPCR21; HG57; hGPCR21; HPN321; KPG_011; PSEC0146
UniProt ID: (Human) Q9NS75
Entrez Gene ID: (Human) 57105
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support