Product Specifications | |
---|---|
Species Reactivity |
Human, Mouse, Rabbit, Rat |
Host/Isotype |
Rabbit / IgG |
Class |
Polyclonal |
Type |
Antibody |
Immunogen |
GST fusion protein with residues 1-46 of rabbit Cav 1.2a, with serine 44 replaced with alanine |
Conjugate |
Unconjugated |
Form |
Lyophilized |
Concentration |
0.95 mg/mL |
Purification |
Antigen affinity chromatography |
Storage buffer |
PBS, pH 7.4, with 1% BSA |
Contains |
0.05% sodium azide |
Storage conditions |
-20°C |
RRID |
AB_2735584 |
Product is shipped at room temperature as a lyophilized powder and should be stored at -20 °C upon receipt. Reconstitution: add 50 µL of deionized water.
CACNA1C encodes an alpha-1 subunit of a voltage-dependent calcium channel. Calcium channels mediate the influx of calcium ions into the cell upon membrane polarization. The alpha-1 subunit consists of 24 transmembrane segments and forms the pore through which ions pass into the cell. The calcium channel consists of a complex of alpha-1, alpha-2/delta, beta, and gamma subunits in a 1:1:1:1 ratio. There are multiple isoforms of each of these proteins, either encoded by different genes or the result of alternative splicing of transcripts. The protein encoded by CACNA1C binds to and is inhibited by dihydropyridine. Alternative splicing results in many transcript variants encoding different proteins.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: brain class C; CaCB receptor; CACB-receptor; calcium channel voltage-dependent alpha1c subunit; calcium channel, cardic dihydropyridine-sensitive, alpha-1 subunit; Calcium channel, L type, alpha-1 polypeptide, isoform 1, cardiac muscle; calcium channel, voltage-dependent, alpha 1C subunit; calcium channel, voltage-dependent, L type, alpha 1C subunit; Cav1.2a; DHPR, alpha-1 subunit; L-type calcium channel alpha-1 subunit; MBC; MELC-CC; Mouse brain class C; neuronal voltage-gated calcium channel alpha 1C subunit; Rat brain class C; RBC; skeletal muscle-specific calcium channel; smooth muscle calcium channel blocker; Smooth muscle calcium channel blocker receptor; unnamed protein product; voltage-dependent L-type Ca2+ channel alpha 1 subunit; Voltage-dependent L-type calcium channel subunit alpha-1C; Voltage-gated calcium channel subunit alpha Cav1.2; voltage-gated L-type calcium channel Cav1.2 alpha 1 subunit, splice variant 10*
Gene Aliases: CACB; CACH2; CACN2; CACNA1C; CACNL1A1; CaV1.2; CCHL1A1; D930026N18Rik; LQT8; MBC; MELC-CC; RATIVS302; TS
UniProt ID: (Human) Q13936, (Rabbit) P15381, (Mouse) Q01815, (Rat) P22002
Entrez Gene ID: (Human) 775, (Rabbit) 100101555, (Mouse) 12288, (Rat) 24239
Molecular Function:
voltage-gated ion channel
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support