Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 1
Immunogen sequence: TLKGLTPTGM LPSGVLSGGK QTLQSATVEA IEADEAIKGF SPQHKITSFE EAKGLDRINE RMPPRRDAMP SDANLNSINK ALTSETNGTD SNGSNSSNIQ
Calcineurin plays an important role in the T-cell receptor-mediated signal transduction pathway. The protein encoded by this gene binds specifically to the activated form of calcineurin and inhibits calcineurin-mediated signal transduction. The encoded protein is found in the nucleus and contains a leucine zipper domain as well as several PEST motifs, sequences which confer targeted degradation to those proteins which contain them. Alternative splicing results in multiple transcript variants encoding two different isoforms.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Calcineurin A alpha; Calcineurin subunit A alpha; CalcineurinA; Calmodulin-dependent calcineurin A subunit alpha isoform; CALNA 1; CAM-PRP catalytic subunit; CCN 1; CNA alpha; PP2B alpha 1; PP2BA alpha; PP2CB; protein phosphatase 2B, catalytic subunit, alpha isoform; protein phosphatase 3 (formerly 2B), catalytic subunit, alpha isoform; Protein phosphatase 3 catalytic subunit alpha; protein phosphatase 3 catalytic subunit alpha isozyme; protein phosphatase 3, catalytic subunit, alpha isoform; protein phosphatase 3, catalytic subunit, alpha isozyme; protein phosphatase-2Ba; Serine/threonine-protein phosphatase 2B catalytic subunit alpha isoform
Gene Aliases: 2900074D19Rik; CALN; CALNA; CALNA1; CCN1; CN; CNA; CNA1; PPP2B; PPP3CA
UniProt ID: (Human) Q08209, (Mouse) P63328, (Rat) P63329
Entrez Gene ID: (Human) 5530, (Mouse) 19055, (Rat) 24674
Molecular Function: protein phosphatase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support