Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This gene encodes a protein with a markedly acidic C terminus; the basic N-terminus is highly homologous to the N-terminus of a related gene, CNN1. Members of the CNN gene family all contain similar tandemly repeated motifs. This encoded protein is associated with the cytoskeleton but is not involved in contraction.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: calponin 3, acidic; Calponin, acidic isoform; Calponin-3; dJ639P13.2.2 (acidic calponin 3)
Gene Aliases: CNN3
UniProt ID: (Human) Q15417
Entrez Gene ID: (Human) 1266
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support