Invitrogen
MA1-22747 detects Canine Distemper Virus in Virus samples.
MA1-22747 has been successfully used in ELISA procedures.
The MA1-22747 immunogen is derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice.
This antibody recognizes canine Distemper Virus.
Protein Aliases: CDV