Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This antibody does not react with uninfected canine serum.
Purity is >90%.
Recommended pairs for ELISA: MA5-18178 (capture) and MA5-18177 (conjugated, detection).
Canine heartworm is a common parasite of dogs worldwide. Heartworm occurs in about 30 other species as well but rarely affects humans. The infection is commonly transmitted by mosquitoes. Adult heartworms can grow to be up to 10 inches in length and can live for 5-7 years in dogs affecting the heart and lungs.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Dirofilaria; dirofilariasis; Filaria
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support