MA1-7012 detects Canine Heartworm.
MA1-7012 has been successfully used in ELISA, immunofluorescence, and Western blot procedures.
The MA1-7012 immunogen is whole Canine Heartworm.
Canine heartworm is a common parasite of dogs worldwide. Heartworm occurs in about 30 other species as well but rarely affects humans. The infection is commonly transmitted by mosquitoes. Adult heartworms can grow to be up to 10 inches in length and can live for 5-7 years in dogs affecting the heart and lungs.
Protein Aliases: Dirofilaria; dirofilariasis; Filaria