The antibody detects endogenous levels of total Carbonic Anhydrase X / CA10 protein.
This gene encodes a protein that belongs to the carbonic anhydrase family of zinc metalloenzymes, which catalyze the reversible hydration of carbon dioxide in various biological processes. The protein encoded by this gene is an acatalytic member of the alpha-carbonic anhydrase subgroup, and it is thought to play a role in the central nervous system, especially in brain development. Multiple transcript variants encoding the same protein have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CA-RP X; carbonic anhydrase X; Carbonic anhydrase-related protein 10; Carbonic anhydrase-related protein X; CARP X; Cerebral protein 15; cerebral protein-15
Gene Aliases: 2700029L05Rik; BB085816; CA-RPX; CA10; Car10; CARPX; HUCEP-15; UNQ533/PRO1076
UniProt ID: (Human) Q9NS85, (Mouse) P61215
Entrez Gene ID: (Human) 56934, (Mouse) 72605
Molecular Function:
dehydratase
lyase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support