Invitrogen
Carbonic anhydrases (CAs) are a family of zinc metalloenzymes. For background information on the CA family, see MIM 114800.[supplied by OMIM, Mar 2008].
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CA-XIII; Carbonate dehydratase XIII; Carbonic anhydrase 13; Carbonic anhydrase XIII
Gene Aliases: 2310075C21Rik; CA13; Car13; CAXIII
UniProt ID: (Human) Q8N1Q1, (Mouse) Q9D6N1
Entrez Gene ID: (Human) 377677, (Mouse) 71934
Molecular Function:
dehydratase
lyase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support