Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
In Western blots, approximately 10% cross-reactivity with recombinant human (rh) CPB1 is observed and less than 1% cross-reactivity with rhCPA1 and recombinant mouse CPA4 is observed.
Reconstitute at 0.2 mg/mL in sterile PBS.
Three different procarboxypeptidases A and two different procarboxypeptidases B have been isolated. The B1 and B2 forms differ from each other mainly in isoelectric point. Carboxypeptidase B1 is a highly tissue-specific protein and is a useful serum marker for acute pancreatitis and dysfunction of pancreatic transplants. It is not elevated in pancreatic carcinoma.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: carboxypeptidase B
Gene Aliases: 0910001A18Rik; 1810063F02Rik; 2210008M23Rik; AI504870
Entrez Gene ID: (Mouse) 76703
Molecular Function: metalloprotease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support