Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Cathepsin H/K/L/V Antibodies

          Invitrogen

          Cathepsin H/K/L/V Recombinant Rabbit Monoclonal Antibody (5A7)

          View all (5) Cathepsin H/K/L/V antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Cathepsin H/K/L/V Recombinant Rabbit Monoclonal Antibody (5A7)

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (12)
          Cathepsin H/K/L/V Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          Cathepsin H/K/L/V Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 12

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Cathepsin H/K/L/V Antibody (MA572632) in IHC (P)

          Immunohistochemistry analysis of Cathepsin H/K/L/V on paraffin-embedded Mouse Kidney tissue. Antigen retrieval by boiling in sodium citrate buffer (pH 6.0) for 15 min. Samples were incubated with Cathepsin H/K/L/V Recombinant Monoclonal (Product # MA572632) using a dilution at 1:200 overnight at 4°C. Followed by conjugated Goat Anti-Rabbit IgG antibody (Red). DAPI (Blue) was used to stain the cell nuclei. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Cathepsin H/K/L/V Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Cathepsin H/K/L/V Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Cathepsin H/K/L/V Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Cathepsin H/K/L/V Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Cathepsin H/K/L/V Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Cathepsin H/K/L/V Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Cathepsin H/K/L/V Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Cathepsin H/K/L/V Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Cathepsin H/K/L/V Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Cathepsin H/K/L/V Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Cathepsin H/K/L/V Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Cathepsin H/K/L/V Antibody in Immunohistochemistry (Paraffin) (IHC (P))

          Product Details

          MA572632

          Applications
          Tested Dilution
          Publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50-1:200
          -

          Immunohistochemistry (Frozen) (IHC (F))

          1:50-1:200
          -
          Product Specifications

          Species Reactivity

          Human, Mouse

          Host/Isotype

          Rabbit / IgG

          Expression System

          HEK293 cells

          Class

          Recombinant Monoclonal

          Type

          Antibody

          Clone

          5A7

          Immunogen

          KLH conjugated synthetic peptide derived from human Cathepsin L/V/K/H.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.125 mg/mL

          Amount

          12.5 µg

          Purification

          Protein A

          Storage buffer

          TBS, pH 7.4, with 50% glycerol, 1% BSA

          Contains

          0.02% ProClin 300

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Target Information

          The protein encoded by this gene is a lysosomal cysteine proteinase that plays a major role in intracellular protein catabolism. Its substrates include collagen and elastin, as well as alpha-1 protease inhibitor, a major controlling element of neutrophil elastase activity. The encoded protein has been implicated in several pathologic processes, including myofibril necrosis in myopathies and in myocardial ischemia, and in the renal tubular response to proteinuria. This protein, which is a member of the peptidase C1 family, is a dimer composed of disulfide-linked heavy and light chains, both produced from a single protein precursor. Multiple alternatively spliced transcript variants have been found for this gene.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: aleurain; alternatively spliced; Cat H; Cat K; Cat L; Cathepsin B3; Cathepsin BA; Cathepsin K; Cathepsin L1; Cathepsin L2; cathepsin L2, preproprotein; Cathepsin O; cathepsin O1; Cathepsin O2; Cathepsin U; Cathepsin V; Cathepsin X; CTSH; CTSK; CTSL; cysteine protease; furless; hCATL AIII; lysosomal cysteine protease; Major excreted protein; MEP; minisatellite 10q detected by probe MMS10; N-benzoylarginine-beta-naphthylamide hydrolase; nackt; p39 cysteine proteinase; Pro-cathepsin H; Procathepsin L; protease; unnamed protein product

          View more View less

          Gene Aliases: 1190035F06Rik; ACC-4; ACC-5; ACC4; ACC5; AI323530; AL022844; catK; CATL; CATL2; CPSB; CTS02; CTSH; CTSK; CTSL; CTSL1; CTSL2; CTSO; CTSO1; CTSO2; CTSU; CTSV; fs; MEP; MMS10-Q; Ms10q; nkt; PKND; PYCD; UNQ268/PRO305

          View more View less

          UniProt ID: (Human) P09668, (Human) P43235, (Human) P07711, (Human) O60911, (Mouse) P49935, (Mouse) P55097, (Mouse) P06797

          View more View less

          Entrez Gene ID: (Human) 1512, (Human) 1513, (Human) 1514, (Human) 1515, (Mouse) 13036, (Mouse) 13038, (Mouse) 13039

          View more View less

          Function(s)
          endopeptidase activity aminopeptidase activity cysteine-type endopeptidase activity serine-type endopeptidase activity protein binding peptidase activity cysteine-type peptidase activity cysteine-type endopeptidase activator activity involved in apoptotic process peptidase activator activity involved in apoptotic process hydrolase activity HLA-A specific activating MHC class I receptor activity kininogen binding identical protein binding macromolecular complex binding thyroid hormone binding fibronectin binding collagen binding proteoglycan binding histone binding serpin family protein binding protein complex binding protein self-association cysteine-type carboxypeptidase activity peptide binding cysteine protease
          Process(es)
          metanephros development T cell mediated cytotoxicity adaptive immune response immune response-regulating signaling pathway proteolysis apoptotic process immune response spermatogenesis positive regulation of gene expression positive regulation of epithelial cell migration neuropeptide catabolic process bradykinin catabolic process antigen processing and presentation positive regulation of cell migration zymogen activation protein destabilization response to retinoic acid membrane protein proteolysis surfactant homeostasis proteolysis involved in cellular protein catabolic process dichotomous subdivision of terminal units involved in lung branching ERK1 and ERK2 cascade cellular response to thyroid hormone stimulus lysosomal protein catabolic process response to odorant positive regulation of apoptotic signaling pathway mitophagy thyroid hormone generation extracellular matrix disassembly collagen catabolic process bone resorption negative regulation of cartilage development protein autoprocessing fusion of virus membrane with host plasma membrane receptor-mediated endocytosis of virus by host cell antigen processing and presentation of exogenous peptide antigen via MHC class II enkephalin processing fusion of virus membrane with host endosome membrane CD4-positive, alpha-beta T cell lineage commitment viral entry into host cell antigen processing and presentation of peptide antigen elastin catabolic process macrophage apoptotic process activation of cysteine-type endopeptidase activity involved in apoptotic process positive regulation of cell proliferation positive regulation of peptidase activity negative regulation of apoptotic process positive regulation of angiogenesis regulation of mitophagy positive regulation of protein targeting to mitochondrion cell communication negative regulation of keratinocyte proliferation protein processing hair follicle morphogenesis catagen decidualization epithelial tube branching involved in lung morphogenesis regulation of actin cytoskeleton reorganization
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-6479c7f679-2z78h:80/100.66.76.236:80.
          git-commit: b70c0dceb888abc3b0dcebb5bc3a5b8753e7e816
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.50.0-Offline