Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • CaV1.2 Antibodies

          Antibodies Inc

          Cav1.2 Ca2+ Channel Monoclonal Antibody (N263/31)

          View all (36) CaV1.2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Cav1.2 Ca2+ Channel Monoclonal Antibody (N263/31)

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (4)
          Cav1.2 Ca2+ Channel Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Cav1.2 Ca2+ Channel Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 4

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Cav1.2 Ca2+ Channel Antibody (75-257) in ICC/IF

          TIRF images (top row), super-resolution GSD localization maps (middle row) and binarized images (bottom row) of immunostained CaV1.2 (Product # 75-257, 1:200) channels and β1ARs in representative fixed adult ventricular myocytes isolated from WT mice under control (A) or ISO-stimulated conditions (B). Merged two-channel image showing relative distributions of CaV1.2 and β1ARs is shown in the third column wi... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Cav1.2 Ca2+ Channel Antibody in Immunocytochemistry (ICC/IF)
          Cav1.2 Ca2+ Channel Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Cav1.2 Ca2+ Channel Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Cav1.2 Ca2+ Channel Antibody in Western Blot (WB)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Cav1.2 Ca2+ Channel Monoclonal Antibody (N263/31)

          Product Details

          75-257

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          Assay-dependent
          -

          Immunohistochemistry (Frozen) (IHC (F))

          Assay-dependent
          -

          Immunocytochemistry (ICC/IF)

          1:100
          -

          ELISA (ELISA)

          Assay-dependent
          -
          Product Specifications

          Species Reactivity

          Guinea pig, Human, Mouse, Rabbit, Rat

          Host/Isotype

          Mouse / IgG2b

          Class

          Monoclonal

          Type

          Antibody

          Clone

          N263/31

          Immunogen

          Fusion protein amino acids 808-874 (cytoplasmic loop between repeat II and III) of rat Cav1.2 (accession number Q5S007) produced recombinantly in E. Coli.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated
          • FL490
          • FL550
          • FL594
          • FL650
          • Request custom conjugation

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Protein A

          Storage buffer

          TBS, pH 7.4

          Contains

          0.065% sodium azide

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Dry ice (international)

          Product Specific Information

          For maximum recovery of product, centrifuge the vial prior to removing the cap.

          Target Information

          CACNA1C encodes an alpha-1 subunit of a voltage-dependent calcium channel. Calcium channels mediate the influx of calcium ions into the cell upon membrane polarization. The alpha-1 subunit consists of 24 transmembrane segments and forms the pore through which ions pass into the cell. The calcium channel consists of a complex of alpha-1, alpha-2/delta, beta, and gamma subunits in a 1:1:1:1 ratio. There are multiple isoforms of each of these proteins, either encoded by different genes or the result of alternative splicing of transcripts. The protein encoded by CACNA1C binds to and is inhibited by dihydropyridine. Alternative splicing results in many transcript variants encoding different proteins.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: brain class C; CaCB receptor; CACH3; CACN4; CACNA 1D; CACNA1C intronic transcript 2; CACNA1C intronic transcript 2 (non-protein coding); CACNL1A2; calcium channel voltage-dependent alpha1c subunit; calcium channel, cardic dihydropyridine-sensitive, alpha-1 subunit; calcium channel, L type, alpha-1 polypeptide, isoform 1, cardiac muscle; calcium channel, voltage-dependent, alpha 1C subunit; calcium channel, voltage-dependent, L type, alpha 1C subunit; CaV1.2; DHP receptor; DHPR, alpha-1 subunit; ion channel; L-type calcium channel alpha-1 subunit; L-type voltage-dependent calcium channel alpha-1 subunit; long-NT; LTCC; MGC120730; neuronal voltage-gated calcium channel alpha 1C subunit; RBC; skeletal muscle-specific calcium channel; smooth muscle calcium channel blocker; unnamed protein product; voltage-dependent calcium channel; voltage-dependent L-type Ca2+ channel alpha 1 subunit; voltage-gated calcium channel subunit alpha Cav1.2

          View more View less

          Gene Aliases: CACB; CACH2; CACN2; Cacna1c; CACNA1C-IT2; CACNL1A1; CaV1.2; CCHL1A1; D930026N18Rik; LQT8; MBC; MELC-CC; NEDHLSS; RATIVS302; TS; TS. LQT8

          View more View less

          UniProt ID: (Human) Q13936, (Rabbit) P15381, (Mouse) Q01815, (Guinea pig) O35505, (Rat) P22002

          View more View less

          Entrez Gene ID: (Rabbit) 100144322, (Human) 775, (Rabbit) 100101555, (Mouse) 12288, (Guinea pig) 100135490, (Rat) 24239

          View more View less

          Function(s)
          ion channel activity voltage-gated calcium channel activity protein binding calmodulin binding high voltage-gated calcium channel activity alpha-actinin binding voltage-gated calcium channel activity involved in cardiac muscle cell action potential voltage-gated calcium channel activity involved in AV node cell action potential enzyme binding protein domain specific binding translation initiation factor binding ion channel binding protein phosphatase 2A binding voltage-gated ion channel
          Process(es)
          immune system development ion transport muscle contraction positive regulation of cytosolic calcium ion concentration heart development regulation of cardiac muscle contraction by regulation of the release of sequestered calcium ion embryonic forelimb morphogenesis camera-type eye development positive regulation of adenylate cyclase activity positive regulation of muscle contraction transmembrane transport calcium ion transport into cytosol cardiac conduction calcium ion transmembrane transport via high voltage-gated calcium channel calcium ion transmembrane transport cardiac muscle cell action potential involved in contraction membrane depolarization during cardiac muscle cell action potential membrane depolarization during AV node cell action potential cell communication by electrical coupling involved in cardiac conduction regulation of heart rate by cardiac conduction calcium ion import across plasma membrane regulation of ventricular cardiac muscle cell action potential membrane depolarization during atrial cardiac muscle cell action potential system process calcium ion transport cellular calcium ion homeostasis exocytosis smooth muscle contraction cell communication chemical synaptic transmission adult walking behavior regulation of heart contraction regulation of blood pressure visual learning calcium ion regulated exocytosis regulation of vasoconstriction signaling insulin secretion growth hormone secretion positive regulation of insulin secretion positive regulation of insulin secretion involved in cellular response to glucose stimulus regulation of membrane potential glucose homeostasis regulation of organ growth smooth muscle contraction involved in micturition calcium ion import cell communication involved in cardiac conduction calcium ion transmembrane import into cytosol positive regulation of membrane depolarization positive regulation of calcium ion transmembrane transport via high voltage-gated calcium channel optic nerve development cerebral cortex development corpus callosum development response to nutrient levels axon development response to alcohol cellular response to beta-amyloid
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-7fb9f846c9-xq5gc:80/100.66.76.64:80.
          git-commit: 8207b5afa5c305bd40e0a4f0b9b53fd46f2a8829
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.59.1-2026.09.91-1.0