PA1-25039 is expected to cross react with mouse, Xenopus laevis, and Drosophilia melanogaster due to sequence homology.
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene plays important roles in the determination of centrosome position and segregation, and in the process of microtubule severing. This protein is localized to the centrosome of interphase cells, and redistributes to the region of the spindle poles during mitosis, reflecting the dynamic behavior of the centrosome during the cell cycle.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: calcium binding protein; Caltractin; Caltractin isoform 2; centrin, EF-hand protein, 1; Centrin-1; EF-hand protein; testicular tissue protein Li 37; testis-specific murine centrin
Gene Aliases: Calt; caltractin; CEN1; CETN; CETN1
UniProt ID: (Human) Q12798, (Mouse) P41209
Entrez Gene ID: (Human) 1068, (Mouse) 26369
Molecular Function:
calcium-binding protein
calmodulin
intracellular calcium-sensing protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support