Search
Search
Nordic
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Reconstituted by adding 1.0 mL sterile distilled water, spun down to remove insoluble particles, divided into small aliquots, frozen and stored at or below -20°C.
Cross-reactivities against enzymes of other sources may occur but have not been determined.
Prior to use, an aliquot is thawed slowly at ambient temperature, spun down again and used to prepare working dilutions by adding sterile phosphate buffered saline (PBS, pH 7.2). Repeated thawing and freezing should be avoided. Working dilutions should be stored at 4°C, not refrozen, and preferably used the same day. If a slight precipitation occurs upon storage, this should be removed by centrifugation. It will not affect the performance of the product.
Ceruloplasmin encodes a protein that is a metalloprotein that binds most of the copper in plasma and is involved in the peroxidation of Fe(II)transferrin to Fe(III) transferrin. Mutations in this gene cause aceruloplasminemia, which results in iron accumulation and tissue damage, and is associated with diabetes and neurologic abnormalities.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: caeruloplasmin; Ceruloplasmin; ceruloplasmin (ferroxidase); Cuproxidase ceruloplasmin; Ferroxidase ceruloplasmin; Glutathione peroxidase ceruloplasmin; Glutathione-dependent peroxiredoxin ceruloplasmin; preceruloplasmin (EC 1.16.3.1); unnamed protein product
Gene Aliases: AB073614; CP; CP-2
UniProt ID: (Human) P00450
Entrez Gene ID: (Human) 1356
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support