Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: IGPLIICKKD SLDKEKEKHI DREFVVMFSV VDENFSWYLE DNIKTYCSEP EKVDKDNEDF QESNRMYSVN GYTFGSLPGL SMCAEDRVKW YLFGMGNEVD VHAAFFHGQA LTNKNYRIDT INLFPATLFD AYMVAQNPGE WMLSCQNLNH LKAGLQAFFQ VQECNKSSSK DNIRGKHVRH YYIAAEEIIW NYAPSGIDIF TKENLTAPGS DSAVFFEQGT TRIGGSYKKL VYREYTDASF TNRKERGPEE EHL
Ceruloplasmin encodes a protein that is a metalloprotein that binds most of the copper in plasma and is involved in the peroxidation of Fe(II)transferrin to Fe(III) transferrin. Mutations in this gene cause aceruloplasminemia, which results in iron accumulation and tissue damage, and is associated with diabetes and neurologic abnormalities.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Ceruloplasmin; Cuproxidase ceruloplasmin; Ferroxidase ceruloplasmin; Glutathione peroxidase ceruloplasmin; Glutathione-dependent peroxiredoxin ceruloplasmin
Gene Aliases: CP; CP-2
UniProt ID: (Human) P00450
Entrez Gene ID: (Human) 1356
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support