NeoBiotechnologies
Applications | Tested Dilution | Publications |
---|---|---|
Immunohistochemistry (Paraffin) (IHC (P)) |
0.25-0.5 µg/mL | - |
Peptide Array (Array) |
Assay-dependent | - |
Product Specifications | |
---|---|
Species Reactivity |
Human, Mouse, Rat |
Host/Isotype |
Mouse / IgG1, kappa |
Class |
Monoclonal |
Type |
Antibody |
Clone |
CHGA, 798 |
Immunogen |
Recombinant full-length human CHGA protein |
Conjugate |
Unconjugated |
Form |
Liquid |
Concentration |
200 µg/mL |
Purification |
Protein G |
Storage buffer |
PBS, pH 7.4, with 0.05% BSA |
Contains |
0.05% sodium azide |
Storage conditions |
4° C |
Immunohistochemistry (PFA fixed): incubate antibody for 30 min at RT. Staining of formalin-fixed tissues requires heating tissue sections in 10mM Tris with 1mM EDTA, pH 9.0, for 45 min at 95 degrees C followed by cooling at RT for 20 minutes.
Chromogranin A (CHGA) is a member of the chromogranin/secretogranin family of neuroendocrine secretory proteins. It is found in secretory vesicles of neurons and endocine cells. CHGA is a precursor to three biologically active peptides: vasostatin, pancreastatin, and parastatin. These peptides act as autocine or paracrine negative modulators of the neuroendocrine system. CHGA is an excellent marker for carcinoid tumors, pheo-chromocytomas, paragangliomas, and other neuro-endocrine tumors. Coexpression of chromogranin A and neuron specific enolase (NSE) is common in neuroendocrine neoplasms.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AL-11; AL26; Beta-granin; betagranin (N-terminal fragment of chromogranin A); Catestatin; CgA; chromofungin; Chromogranin-A; CTNNB; DKFZp686D02253; EA-92; ER-37; ES-43; FLJ25606; FLJ37923; GE-25; GR-44; GV-19; LF-19; MGC105435; p-Glu serpinin precursor; Pancreastatin; parathyroid secretory protein 1; Pituitary secretory protein I; Serpinin; Serpinin-RRG; SL21; SP-I; SS-18; Vasostatin I; Vasostatin II; Vasostatin-1; Vasostatin-2; WA-8; WE-14
Gene Aliases: CGA; CHGA; ChrA
UniProt ID: (Human) P10645, (Mouse) P26339
Entrez Gene ID: (Human) 1113, (Rat) 24258, (Mouse) 12652
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support