Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
MA1-91500 detects Chymotrypsin from human samples.
MA1-91500 has been successfully used in ELISA and Western blot applications.
The MA1-91500 immunogen is: Purified Pancreatic Chymotrypsin (Human).
This gene encodes a member of the peptidase S1 family. The encoded protein is a serum calcium-decreasing factor that has chymotrypsin-like protease activity. Alternatively spliced transcript variants have been observed, but their full-length nature has not been determined.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Caldecrin; chymotrypsin C (caldecrin); Chymotrypsin-C; elastase 4; elastase IV; serum calcium decreasing factor
Gene Aliases: CLCR; CTRC
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support