Applications | Tested Dilution | Publications |
---|---|---|
Western Blot (WB) |
1:1,000-1:5,000 | - |
Immunoprecipitation (IP) |
Assay-dependent | - |
Product Specifications | |
---|---|
Species Reactivity |
Human |
Host/Isotype |
Rabbit / IgG |
Class |
Polyclonal |
Type |
Antibody |
Immunogen |
Between 450 and 500 |
Conjugate |
Unconjugated |
Form |
liquid |
Concentration |
1 mg/mL |
Storage conditions |
4° C |
The recommended shelf life for this product is 1 year from date of receipt.
Xenopus claspin is an essential upstream regulator of checkpoint kinase 1 and triggers a checkpoint arrest of the cell cycle in the presence of DNA templates in Xenopus egg extracts. The human gene appears to be the homolog Xenopus claspin and its function has not been determined.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Claspin; claspin homolog; hClaspin; MGC131612; MGC131613; MGC131615
Gene Aliases: CLSPN
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support