Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
PA1-28518 detects Clathrin from human, bovine samples.
PA1-28518 has been successfully used in immunocytochemistry applications.
The PA1-28518 immunogen is full length native protein (purified) (Cow)
AP-1 complex subunit beta-1 is a subunit of clathrin-associated adaptor protein complex 1 that plays a role in protein sorting in the late-Golgi/trans-Golgi network (TGN) and/or endosomes. The AP complexes mediate both the recruitment of clathrin to membranes and the recognition of sorting signals within the cytosolic tails of transmembrane cargo molecules.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: adapter-related protein complex 1 subunit beta-1; Adaptor protein complex AP-1 subunit beta-1; adaptor related protein complex 1 beta 1 subunit; adaptor-related protein complex 1 beta 1 subunit; Adaptor-related protein complex 1 subunit beta-1; adaptor-related protein complex 1, beta 1 subunit; ADTB2; AP-1 complex subunit beta-1; AP2B1; Beta-1-adaptin; Beta-adaptin 1; beta-prime-adaptin; beta1-adaptin; CLAPB1; Clathrin assembly protein complex 1 beta large chain; Golgi adaptor HA1/AP1 adaptin beta subunit; plasma membrane adaptor HA2/AP2 adaptor beta subunit
Gene Aliases: ADTB1; AP1B1; BAM22; CLAPB2
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support