Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
MA1-25736 detects Clathrin light chain from bovine samples. MA1-25736 is expected to cross react with human (100% conserved), mouse (100% conserved), rat (100% conserved), Xenopus laevis (100% conserved), and zebrafish (95% conserved) due to sequence homo
MA1-25736 has been successfully used in immunoprecipitation, and Western blot procedures.
The MA1-25736 immunogen is a synthetic peptide corresponding to residues E(23) E D P A A A F L A Q Q E S E I A G I E N D(44) of the conserved regions of bovine Clathrin Light Chain.
Clathrin is the major protein of the polyhedral coat of coated pits and vesicles. Two different adapter protein complexes link the clathrin lattice either to the plasma membrane or to the trans-Golgi network.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CHC17; Clathrin light chain A; Clathrin light chain B; clathrin, light chain (Lca); clathrin, light chain (Lcb); clathrin, light chain A; clathrin, light chain A2; clathrin, light chain B; clathrin, light polypeptide (Lca); clathrin, light polypeptide (Lcb); clathrin, light polypeptide A b; clathrin, light polypeptide B (light chain B); clathryn light chain (LCA2); CLH17; CLTC; CTLA; CTLB; KIAA0034; Lca; Lcb; unnamed protein product
Gene Aliases: CLTA; CLTB; CLTLB
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support