Invitrogen
PA5-16875 targets Claudin 4 in IHC (P) applications and shows reactivity with Human and mouse samples.
The PA5-16875 immunogen is a synthetic peptide derived from the C-terminus of human Claudin-4.
Claudin-4 belongs to the 20-member family of claudins, transmembrane cellular adhesion proteins localized at epithelial and endothelial tight junctions. Claudins demonstrate distinct, tissue-specific expression patterns, and are essential components of tight junction charge-specific channels that regulate paracellular ion flux. Claudin-4 is a component of tight junction strands and may play a role in internal organ development and function during pre- and postnatal life. The overexpression of Claudin-4 has been shown to decrease paracellular electrical conductance due to a selective decrease in Na+ permeability, with no significant decrease in Cl- permeability. In ovarian carcinoma cells, Claudin-3 and Claudin-4 exhibit upregulation at both mRNA and protein levels. Claudin-4 is also overexpressed in pancreatic cancer tissues and cell lines, and in other epithelial cancers. The Claudin-4 gene is deleted in Williams-Beuren syndrome, a neurodevelopmental disorder affecting multiple systems.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Claudin-4; CLAUDIN4; Clostridium perfringens enterotoxin receptor; Clostridium perfringens enterotoxin receptor 1; CPE-R; CPE-receptor; h CPE-receptor; Williams-Beuren syndrome chromosomal region 8 protein
Gene Aliases: Cep-r; CLDN4; CPE-R; CPER; CPETR; CPETR1; hCPE-R; WBSCR8
UniProt ID: (Human) O14493, (Mouse) O35054
Entrez Gene ID: (Human) 1364, (Mouse) 12740
Molecular Function:
cell junction protein
tight junction
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support