Recommended positive controls: 293T, HepG2.
Predicted reactivity: Cow: 85%; Dog: 77%; Horse: 85%; Pig: 77%; Rabbit: 85%; Rat: 77%.
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Claudin-8 belongs to the claudin family of integral membrane proteins. Claudins are responsible for the formation of tight junction strands, and are connected with the actin cytoskeleton mediated by ZO-1. Claudin-8 is a tight junction protein expressed in several distal nephron segments, suggesting that it may have a role as a paracellular cation barrier protein. Claudin-8 is expressed in the inner ear, and the umbrella cell tight junctions in the bladder epithelium.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Claudin-8; epididymis secretory protein Li 79
Gene Aliases: AI648025; CLDN8; HEL-S-79; UNQ779/PRO1573
UniProt ID: (Human) P56748, (Mouse) Q9Z260
Entrez Gene ID: (Human) 9073, (Mouse) 54420
Molecular Function:
cell junction protein
tight junction
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support