Invitrogen
The MA1-7411 antibody reacts with Clostridium botulinum D toxoid in bacterial and infected tissue samples.
MA1-7411 has been successfully used in ELISA and Immunofluorescence applications.
The MA1-7411 antibody was raised against Clostridium botulimum D toxoid.
Clostridium botulinum is a gram-positive, anaerobic motile bacteria normally found in soil. C. botulinum is a potential threat to both humans and animals through contaminated food because of the potent neurotoxin they produce. C. botulinum bacteria are categorized within 7 strains A-G based on the characteristics of their toxin. A,B, E and F strains are known to cause food poisoning in humans.
Protein Aliases: C. botulinum Toxin D; C. botulinum Toxoid D