Invitrogen
The MA1-7416 antibody reacts with Clostridium difficile toxin A in bacterial and infected tissue samples.
MA1-7416 has been successfully used in ELISA applications.
The MA1-7416 antibody was raised against Toxina A.
Clostridium difficile is a species of bacteria of the genus Clostridium which are Gram-positive, anaerobic, spore-forming rods (bacillus). C.difficile is the most significant cause of pseudomembranous colitis, a severe infection of the colon.
Protein Aliases: TcdA